2-Chloro-5-methylnicotinic acid

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-methylpyridine-3-carboxylic acid

054072-250MG 054072-1G 054072-5G
102833-184EA 300 USD
102833-184 102833-186 102833-188
2-Chloro-5-methylnicotinic acid
2-Chloro-5-methylnicotinic acid

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.5800 g/mol
MDL Number: MFCD07375371
CAS Number: 66909-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR