β-Alaninamide hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-Aminopropionamide Hydrochloride, H-β-Ala-NH₂·HCl, H-β-Ala-NH2.HCl, H-β-Ala-NH2·HCl

052750-5G 052750-25G
102833-176EA 32.31 USD
102833-176 102833-178
β-Alaninamide hydrochloride
beta-Alaninamide hydrochloride

Hazard: Irritant

Formula: C₃H₈N₂O·HCl
MW: 124.57 g/mol
MDL Number: MFCD00060747
CAS Number: 64017-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR