[2-(1-Piperidinyl)phenyl]methanamine
Supplier: Matrix Scientific
Synonyms:
2-(Piperidin-1-yl)benzylamine
052586-250MG
052586-1G
102551-396EA
66.15
USD
102551-396
102551-398
[2-(1-Piperidinyl)phenyl]methanamine
[2-(1-Piperidinyl)phenyl]methanamine
Hazard: Irritant
Formula:
C₁₂H₁₈N₂ |
MDL Number:
MFCD02682071 CAS Number: 72752-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...