4-(6-Chloropyrazin-2-yl)morpholine

Supplier: Matrix Scientific

051890-1G 051890-5G
102833-170EA 267.69 USD
102833-170 102833-172
4-(6-Chloropyrazin-2-yl)morpholine
4-(6-Chloropyrazin-2-yl)morpholine

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
CAS Number: 720693-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR