3-Methoxyazetidine hydrochloride

Supplier: Matrix Scientific

049460-1G 049460-5G
102551-388EA 21.54 USD
102551-388 102551-390
3-Methoxyazetidine hydrochloride
3-Methoxyazetidine hydrochloride

Hazard: Irritant

Formula: C4H10ClNO
MW: 123.58 g/mol
MDL Number: MFCD06804514
CAS Number: 148644-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR