Ac-DL-Pro-OH

Supplier: Matrix Scientific

Synonyms: 1-Acetylpyrrolidine-2-carboxylic acid, N-Acetyl-DL-Proline

049052-5G 049052-25G
102833-166EA 95.38 USD
102833-166 102833-168
Ac-DL-Pro-OH
Ac-DL-Pro-OH

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD00063230
CAS Number: 1074-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR