3-Bromo-4-methylbenzamide
Supplier: Matrix Scientific
048760-5G
048760-25G
102833-162EA
212.31
USD
102833-162
102833-164
3-Bromo-4-methylbenzamide
3-Bromo-4-methylbenzamide
Hazard: Irritant
Formula:
C₈H₈BrNO MW: 214.0592 g/mol |
MDL Number:
MFCD03306087 CAS Number: 183723-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...