Gabapentin ≥98%
Supplier: Matrix Scientific
Synonyms:
2-[1-(Aminomethyl)cyclohexyl]acetic acid, Neurontin
047892-25G
047892-100G
102833-154EA
53.85
USD
102833-154
102833-156
Gabapentin ≥98%
Gabapentin
Hazard: Irritant
Formula:
C₉H₁₇NO₂ MW: 171.24 g/mol |
MDL Number:
MFCD00865286 CAS Number: 60142-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...