3-Chloro-5-(trifluoromethyl)picolinic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-5-(trifluoromethyl)-2-pyridinecarboxylic acid, 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 3-Chloro-5-(trifluoromethyl)picolinic acid
046766-1G
046766-5G
102518-842EA
38.46
USD
102518-842
102518-844
3-Chloro-5-(trifluoromethyl)picolinic acid ≥95%
3-Chloro-5-(trifluoromethyl)picolinic acid
Hazard: Irritant
Formula:
C₇H₃ClF₃NO₂ MW: 225.60 g/mol Melting Pt: 141…143 °C |
MDL Number:
MFCD00277482 CAS Number: 80194-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...