N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
Supplier: Matrix Scientific
Synonyms:
N-Fmoc-D-alanine, Fmoc-D-Ala-OH
041256-25G
041256-100G
102551-280EA
110.77
USD
102551-280
102663-700
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
Hazard: Irritant
Formula:
C₁₈H₁₇NO₄ |
MDL Number:
MFCD00062960 CAS Number: 79990-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...