3-(Chloromethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Cyanobenzyl chloride

041234-5G 041234-25G
102663-696EA 20 USD
102663-696 102663-698
3-(Chloromethyl)benzonitrile ≥97%
3-(Chloromethyl)benzonitrile

Hazard: Irritant

Formula: C₈H₆ClN
Melting Pt: 66…71 °C
MDL Number: MFCD02684191
CAS Number: 64407-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR