Ethyl-5-amino-1H-pyrazole-3-carboxylate
Supplier: Matrix Scientific
Synonyms:
Ethyl 5-amino-1H-pyrazole-3-carboxylate
040879-1G
040879-5G
102833-106EA
129.23
USD
102833-106
102833-108
Ethyl-5-amino-1H-pyrazole-3-carboxylate
Ethyl-5-amino-1H-pyrazole-3-carboxylate
Hazard: Irritant
Formula:
C₆H₉N₃O₂ MW: 155.1600 g/mol |
MDL Number:
MFCD03468385 CAS Number: 105434-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...