1H-Benzimidazole-5-carboxylic acid
Supplier: Matrix Scientific
039498-1G
039498-5G
039498-25G
102551-266EA
15.38
USD
102551-266
102551-268
102551-270
1H-Benzimidazole-5-carboxylic acid
1H-Benzimidazole-5-carboxylic acid
Hazard: Irritant
Formula:
C₈H₆N₂O₂ |
MDL Number:
MFCD00011555 CAS Number: 15788-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...