2-Ethoxyphenyl isocyanate

Supplier: Matrix Scientific

Synonyms: 1-Ethoxy-2-isocyanatobenzene

038317-5G 038317-25G
102833-088EA 143.08 USD
102833-088 102833-090
2-Ethoxyphenyl isocyanate
2-Ethoxyphenyl isocyanate

Hazard: Irritant

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00002007
CAS Number: 5395-71-1
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR