2,5-Dimethoxyphenyl isocyanate

Supplier: Matrix Scientific

038310-5G 038310-25G
102551-256EA 93.85 USD
102551-256 102551-258
2,5-Dimethoxyphenyl isocyanate
2,5-Dimethoxyphenyl isocyanate

Hazard: Toxic/Refrigerate

Formula: C₉H₉NO₃
Melting Pt: 40…42 °C
MDL Number: MFCD00002006
CAS Number: 56309-62-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR