3-Amino-3-(4-bromophenyl)propionic acid

Supplier: Matrix Scientific

037905-1G 037905-5G
102551-246EA 207.69 USD
102551-246 102551-248
3-Amino-3-(4-bromophenyl)propionic acid
3-Amino-3-(4-bromophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₁₀BrNO₂
MW: 244.0900 g/mol
MDL Number: MFCD00187112
CAS Number: 39773-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR