1-Benzylpiperidine-4-carbaldehyde

Supplier: Matrix Scientific

Synonyms: N-Benzylpiperidine-4-carboxaldehyde, 1-Benzyl-4-piperidinecarboxaldehyde

037718-5G 037718-25G
102551-242EA 126.15 USD
102551-242 102551-244
1-Benzylpiperidine-4-carbaldehyde
1-Benzylpiperidine-4-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₇NO
MW: 203.28 g/mol
MDL Number: MFCD02258902
CAS Number: 22065-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR