4-Hydroxy-3-methoxybenzylamine hydrochloride

Supplier: Matrix Scientific

Synonyms: Vanillylamine HCl, 4-(Aminomethyl)-2-methoxyphenol hydrochloride

037616-5G 037616-25G 037616-100G
102551-232EA 15.38 USD
102551-232 102551-234 102551-236
4-Hydroxy-3-methoxybenzylamine hydrochloride
4-Hydroxy-3-methoxybenzylamine hydrochloride

Hazard: Irritant

Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Melting Pt: 227°C
MDL Number: MFCD00012864
CAS Number: 7149-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR