5-Methyl-1H-pyrazole ≥95%
Supplier: Matrix Scientific
037514-5G
037514-25G
037514-100G
102551-218EA
15.38
USD
102551-218
102551-220
102551-222
5-Methyl-1H-pyrazole ≥95%
5-Methyl-1H-pyrazole
Hazard: Irritant
Formula:
C₄H₆N₂ |
MDL Number:
MFCD08685900 CAS Number: 88054-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...