3-(2-Aminoethyl)pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Pyridin-3-yl)ethanamine, 2-(3-Pyridyl)ethylamine
037494-1G
037494-5G
102833-074EA
32.31
USD
102833-074
102833-076
3-(2-Aminoethyl)pyridine ≥95%
3-(2-Aminoethyl)pyridine
Hazard: Corrosive/Air Sensitive
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Boiling Pt: 119-120°C/19mm |
MDL Number:
MFCD00191604 CAS Number: 20173-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...