3-Acetylthiophene
Supplier: Matrix Scientific
Synonyms:
Methyl 3-thienyl ketone, 1-(3-Thienyl)-1-ethanone
037464-5G
037464-25G
102833-064EA
43.08
USD
102833-064
102833-066
3-Acetylthiophene
3-Acetylthiophene
Hazard: Irritant
Formula:
C₆H₆OS MW: 126.18 g/mol |
MDL Number:
MFCD00005468 CAS Number: 1468-83-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...