3-Methylbenzylamine

Supplier: Matrix Scientific

Synonyms: m-Tolylmethanamine, 3-(Aminomethyl)toluene

037449-5G 037449-25G
102551-214EA 89.23 USD
102551-214 102551-216
3-Methylbenzylamine
3-Methylbenzylamine

Hazard: Irritant

Formula: C₈H₁₁N
MW: 121.18 g/mol
MDL Number: MFCD00008118
CAS Number: 100-81-2
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR