4-Amino-2-chloro-6,7-dimethoxyquinazoline
Supplier: Matrix Scientific
Synonyms:
Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline
037434-5G
037434-25G
102833-060EA
93.85
USD
102833-060
102833-062
4-Amino-2-chloro-6,7-dimethoxyquinazoline
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Hazard: Irritant
Formula:
C₁₀H₁₀ClN₃O₂ MW: 239.66 g/mol |
MDL Number:
MFCD00051734 CAS Number: 23680-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...