L-Histidine methyl ester dihydrochloride

Supplier: Matrix Scientific

Synonyms: L(+)-Histidine methyl ester dihydrochloride, H-His-OMe.2HCl, L-Methyl histidinate dihydrochloride, H-His-OMe·2HCl

037418-25G 037418-100G
102551-210EA 103.08 USD
102551-210 102551-212
L-Histidine methyl ester dihydrochloride
L-Histidine methyl ester dihydrochloride

Hazard: Irritant

Formula: C₇H₁₁N₃O₂
MDL Number: MFCD00012701
CAS Number: 7389-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR