5-Nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

Supplier: Matrix Scientific

037131-5G 037131-25G
102551-198EA 300 USD
102551-198 102551-200
5-Nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
5-Nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₃N₃O₆
MDL Number: MFCD00102613
CAS Number: 17687-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR