4-Amino-2-nitrobenzoic acid

Supplier: Matrix Scientific

037041-1G 037041-5G
102551-192EA 47.69 USD
102551-192 102551-194
4-Amino-2-nitrobenzoic acid
4-Amino-2-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₆N₂O₄
MDL Number: MFCD00189376
CAS Number: 610-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR