3,5-Dimethyl-4-isoxazolecarbaldehyde

Supplier: Matrix Scientific

Synonyms: 3,5-Dimethylisoxazole-4-carboxaldehyde, 3,5-Dimethylisoxazol-4-carboxaldehyde

036452-1G 036452-5G
102551-182EA 18.46 USD
102551-182 102551-184
3,5-Dimethyl-4-isoxazolecarbaldehyde
3,5-Dimethyl-4-isoxazolecarbaldehyde

Hazard: Irritant

Formula: C6H7NO2
MW: 125.13 g/mol
MDL Number: MFCD02681977
CAS Number: 54593-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR