Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 4-Quinazolone-2-carboxylate, 4-Quinazolone-2-carboxylic acid ethyl ester

036002-1G 036002-5G
102551-174EA 123.08 USD
102551-174 102551-176
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD00051583
CAS Number: 29113-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR