5-Bromo-3-methoxypicolinic acid

Supplier: Matrix Scientific

035700-250MG 035700-1G
102833-042EA 500 USD
102833-042 102833-044
5-Bromo-3-methoxypicolinic acid
5-Bromo-3-methoxypicolinic acid

Hazard: Irritant

Formula: C₇H₆BrNO₃
MW: 232.0300 g/mol
MDL Number: MFCD11857696
CAS Number: 1142191-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR