5-Acetyl-2-fluorophenylboronic acid
Supplier: Matrix Scientific
Synonyms:
(5-Acetyl-2-fluorophenyl)boronic acid
035626-1G
035626-5G
102518-830EA
78.46
USD
102518-830
102518-832
5-Acetyl-2-fluorophenylboronic acid
5-Acetyl-2-fluorophenylboronic acid
Hazard: Irritant
Formula:
C₈H₈BFO₃ |
MDL Number:
MFCD06656263 CAS Number: 870777-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...