4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Supplier: Matrix Scientific
Synonyms:
4-Fluoro-2-(trifluoromethyl)benzeneboronic acid, (4-Fluoro-2-(trifluoromethyl)phenyl)boronic acid
035596-5G
035596-25G
102833-034EA
240
USD
102833-034
102833-036
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Hazard: Irritant
Formula:
C₇H₅BF₄O₂ MW: 207.92 g/mol |
MDL Number:
MFCD08062379 CAS Number: 182344-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...