4-Ethoxy-2,6-difluorophenylboronic acid
Supplier: Matrix Scientific
Synonyms:
2,6-Difluoro-4-ethoxyphenylboronic acid, (4-Ethoxy-2,6-difluorophenyl)boronic acid
035582-1G
035582-5G
102833-030EA
367.69
USD
102833-030
102833-032
4-Ethoxy-2,6-difluorophenylboronic acid
4-Ethoxy-2,6-difluorophenylboronic acid
Hazard: Irritant/Refrigerate
Formula:
C₈H₉BF₂O₃ MW: 201.97 g/mol |
MDL Number:
MFCD12026735 CAS Number: 1310403-94-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...