4-Chloro-2-methylphenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-methylbenzeneboronic acid

035573-5G 035573-25G
102518-824EA 26.15 USD
102518-824 102518-826
4-Chloro-2-methylphenylboronic acid
4-Chloro-2-methylphenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₈BClO₂
MW: 170.40 g/mol
Melting Pt: 193-195°C
MDL Number: MFCD02683107
CAS Number: 209919-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR