3-Cyano-4-fluorophenylboronic acid
Supplier: Matrix Scientific
Synonyms:
3-Cyano-4-fluorobenzeneboronic acid
035515-1G
035515-5G
102663-678EA
126.15
USD
102663-678
102663-680
3-Cyano-4-fluorophenylboronic acid
3-Cyano-4-fluorophenylboronic acid
Hazard: Irritant
Formula:
C₇H₅BFNO₂ MW: 164.93 g/mol |
MDL Number:
MFCD03095130 CAS Number: 214210-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...