2-Ethylphenylboronic acid
Supplier: Matrix Scientific
Synonyms:
(2-Ethylphenyl)metaboric acid, 2-Ethylbenzeneboronic acid, Dihydroxy(2-ethylphenyl)borane
035422-5G
035422-25G
102833-020EA
36.92
USD
102833-020
102833-022
2-Ethylphenylboronic acid
2-Ethylphenylboronic acid
Hazard: Irritant/Store in Freezer
Formula:
C₈H₁₁BO₂ MW: 149.98 g/mol Melting Pt: 96-98°C |
MDL Number:
MFCD02093075 CAS Number: 90002-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...