2-Chloro-5-formylphenylboronic acid

Supplier: Matrix Scientific

Synonyms: (2-Chloro-5-formylphenyl)boronic acid

035396-1G 035396-5G
102518-816EA 138.46 USD
102518-816 102518-818
2-Chloro-5-formylphenylboronic acid
2-Chloro-5-formylphenylboronic acid

Hazard: Irritant

Formula: C₇H₆BClO₃
MW: 184.3847 g/mol
MDL Number: MFCD06656269
CAS Number: 1150114-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR