2,6-Dimethylphenylboronic acid

Supplier: Matrix Scientific

Synonyms: 2,6-Dimethylbenzeneboronic acid, (2,6-Dimethylphenyl)boronic acid, Dihydroxy(2,6-dimethylphenyl)borane, (2,6-Dimethylphenyl)metaboric acid

035326-5G 035326-25G
102518-812EA 44.62 USD
102518-812 102518-814
2,6-Dimethylphenylboronic acid
2,6-Dimethylphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 120……124 °C
MDL Number: MFCD01009693
CAS Number: 100379-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR