2,5-Dimethylphenylboronic acid

Supplier: Matrix Scientific

035316-5G 035316-25G
102833-012EA 21.54 USD
102833-012 102833-014
2,5-Dimethylphenylboronic acid
2,5-Dimethylphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₈H₁₁BO₂
Melting Pt: 186…191 °C
MDL Number: MFCD01863525
CAS Number: 85199-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR