2,3-Difluoro-4-ethoxyphenylboronic acid
Supplier: Matrix Scientific
035291-1G
035291-5G
035291-25G
102833-006EA
36.92
USD
102833-006
102833-008
102833-010
2,3-Difluoro-4-ethoxyphenylboronic acid
2,3-Difluoro-4-ethoxyphenylboronic acid
Hazard: Irritant
Formula:
C₈H₉BF₂O₃ Melting Pt: 151…154 °C |
MDL Number:
MFCD09258743 CAS Number: 212386-71-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...