2-Iodo-3-pyridinol
Supplier: Matrix Scientific
Synonyms:
3-Hydroxy-2-iodopyridine
035137-5G
035137-25G
102833-002EA
95.38
USD
102833-002
102833-004
2-Iodo-3-pyridinol
2-Iodo-3-pyridinol
Hazard: Irritant
Formula:
C₅H₄INO MW: 221.00 g/mol |
MDL Number:
MFCD00023421 CAS Number: 40263-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...