(S)-2-Amino-3-phenylpropanamide
Supplier: Matrix Scientific
Synonyms:
H-Phe-NH₂, L-Phenylalaninamide
034254-5G
034254-25G
102832-998EA
118.46
USD
102832-998
102833-000
(S)-2-Amino-3-phenylpropanamide
(S)-2-Amino-3-phenylpropanamide
Hazard: Irritant
Formula:
C₉H₁₂N₂O MW: 164.20 g/mol |
MDL Number:
MFCD00038146 CAS Number: 5241-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...