4-Iodo-1-methyl-1H-pyrazole
Supplier: Matrix Scientific
034053-5G
034053-25G
102663-666EA
21.54
USD
102663-666
102663-668
4-Iodo-1-methyl-1H-pyrazole
4-Iodo-1-methyl-1H-pyrazole
Hazard: Irritant
Formula:
C₄H₅IN₂ MW: 208.0000 g/mol Melting Pt: 59…64 °C |
MDL Number:
MFCD05663860 CAS Number: 39806-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...