Benzofuran-2-carboxaldehyde

Supplier: Matrix Scientific

Synonyms: Benzo[b]furan-2-carboxaldehyde, Benzofuran-2-carbaldehyde

033801-1G 033801-5G
102551-156EA 158.46 USD
102551-156 102551-158
Benzofuran-2-carboxaldehyde
Benzofuran-2-carboxaldehyde

Hazard: Irritant

Formula: C₉H₆O₂
MW: 146.145 g/mol
MDL Number: MFCD00015463
CAS Number: 4265-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR