2-Bromo-2'-(trifluoromethyl)acetophenone
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[2-(trifluoromethyl)phenyl]-1-ethanone
033216-1G
033216-5G
102551-152EA
98.46
USD
102551-152
102551-154
2-Bromo-2'-(trifluoromethyl)acetophenone
2-Bromo-2'-(trifluoromethyl)acetophenone
Hazard: Irritant
Formula:
C₉H₆BrF₃O MW: 267.04 g/mol |
MDL Number:
MFCD03094304 CAS Number: 54109-16-9 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...