4-Fluororesorcinol ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Fluoro-2,4-dihydroxybenzene

032355-1G 032355-5G
102832-988EA 93.85 USD
102832-988 102832-990
4-Fluororesorcinol ≥97%
4-Fluororesorcinol

Hazard: Irritant/Store in Freezer

Formula: C₆H₅FO₂
MW: 128.10 g/mol
Melting Pt: 96…100 °C
MDL Number: MFCD03789074
CAS Number: 103068-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR