Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Supplier: Matrix Scientific
Synonyms:
Methyl 3-aminothieno[2,3-b]pyridine-2-carboxylate
031045-1G
031045-5G
102518-806EA
255.38
USD
102518-806
102518-808
Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₉H₈N₂O₂S MW: 208.24 g/mol |
MDL Number:
MFCD00174679 CAS Number: 111042-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...