2-Fluoro-4-isocyanato-1-methylbenzene
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-4-methylphenyl isocyanate
030183-1G
030183-5G
030183-25G
102832-976EA
89.23
USD
102832-976
102832-978
102832-980
2-Fluoro-4-isocyanato-1-methylbenzene
2-Fluoro-4-isocyanato-1-methylbenzene
Hazard: Irritant-Harmful/Moisture Sensitive/Refrigerate/Lachrymator/Stored under Argon
Formula:
C₈H₆FNO MW: 151.14 g/mol Boiling Pt: 187…192 °C Flash Pt: 79 °C |
MDL Number:
MFCD00037071 CAS Number: 102561-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...