o-Toluenesulfonyl chloride
Supplier: Matrix Scientific
Synonyms:
Toluene-o-sulphonyl chloride, Toluene-2-sulphonyl chloride, 2-Toluenesulphonyl chloride
028331-5G
028331-25G
102551-118EA
116.92
USD
102551-118
102551-120
o-Toluenesulfonyl chloride
o-Toluenesulphonyl chloride
Hazard: Corrosive/Stored under Argon/Moisture Sensitive/Lachrymator
Formula:
C₇H₇ClO₂S MW: 190.65 g/mol Boiling Pt: 254°C Melting Pt: 10°C Flash Pt: 110°C |
MDL Number:
MFCD00024874 CAS Number: 133-59-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...