1-Methyl-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-Methylpyrazole-3-carboxylic acid

027386-1G 027386-5G
102551-114EA 84.62 USD
102551-114 102551-116
1-Methyl-1H-pyrazole-3-carboxylic acid
1-Methyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
Boiling Pt: 306 °C
Melting Pt: 150…152 °C
Flash Pt: 139 °C
MDL Number: MFCD00464254
CAS Number: 25016-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR