4-Methylthiazole-5-carbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Methylthiazole-5-carboxaldehyde, 5-formyl-4-methyl thiazole, 4-Methyl-5-formylthiazole
027269-1G
027269-5G
102551-106EA
15.38
USD
102551-106
102551-108
4-Methylthiazole-5-carbaldehyde ≥95%
4-Methylthiazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₅H₅NOS MW: 127.161 g/mol |
MDL Number:
MFCD07368277 CAS Number: 82294-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...